Learn More
Medchemexpress LLC (Rac)-IBT6A | 1412418-47-3 | 98.2% | C22H22N6O | 10MG
SDP
Catalog No. 5000260793
Encompass_Preferred
A racemic form of IBT6A, this small-molecule impurity of the BTK inhibitor ibrutinib is supplied for research use as a synthetic intermediate and analytical reference. The solid is characterized by molecular formula C22H22N6O and molecular weight 386.45 g·mol⁻¹, with batch COA confirming typical purity of 98.18% by LCMS.
  • racemic small-molecule impurity useful for synthetic chemistry
  • suitable as an analytical reference or impurity standard
  • characterized by LCMS with typical purity of 98.2%
  • provided as a solid, white to off-white appearance
  • available in milligram-scale quantities for research applications
  • molecular formula C22H22N6O and molecular weight 386.45 g·mol⁻¹
Catalog No. 50-002-60793 Supplier Medchemexpress LLC Supplier No. HY1303610MG
A racemic form of IBT6A, this small-molecule impurity of the BTK inhibitor ibrutinib is supplied for research use as a synthetic intermediate and analytical reference. The solid is characterized by molecular formula C22H22N6O and molecular weight 386.45 g·mol⁻¹, with batch COA confirming typical purity of 98.18% by LCMS.
  • racemic small-molecule impurity useful for synthetic chemistry
  • suitable as an analytical reference or impurity standard
  • characterized by LCMS with typical purity of 98.2%
  • provided as a solid, white to off-white appearance
  • available in milligram-scale quantities for research applications
  • molecular formula C22H22N6O and molecular weight 386.45 g·mol⁻¹