Learn More
Medchemexpress LLC (Rac)-Idroxioleic acid (2-hydroxyoleic acid) | 56472-29-8 | 99.9% | 25 MG
SDP
Catalog No. 5000341595
Encompass_Preferred
(Rac)-Idroxioleic acid (2-Hydroxyoleic acid) is a synthetic oleic acid (OA) derivative. It functions by binding to the plasma membrane and altering lipid organization. This compound has demonstrated anti-tumor effects.
  • Binds to the plasma membrane and alters lipid organization.
  • Exhibits anti-tumor effects.
  • Impairs Jurkat cell growth in a time- and concentration-dependent manner (IC50 of approximately 40 μM).
  • Induces proteolytic cleavage of PARP, a molecular marker of apoptosis.
  • Significantly inhibits tumor growth in nude mice in a xenograft model.
Catalog No. 50-003-41595 Supplier Medchemexpress LLC Supplier No. 25MGHY129467
(Rac)-Idroxioleic acid (2-Hydroxyoleic acid) is a synthetic oleic acid (OA) derivative. It functions by binding to the plasma membrane and altering lipid organization. This compound has demonstrated anti-tumor effects.
  • Binds to the plasma membrane and alters lipid organization.
  • Exhibits anti-tumor effects.
  • Impairs Jurkat cell growth in a time- and concentration-dependent manner (IC50 of approximately 40 μM).
  • Induces proteolytic cleavage of PARP, a molecular marker of apoptosis.
  • Significantly inhibits tumor growth in nude mice in a xenograft model.