Learn More
Medchemexpress LLC (Rac)-Idroxioleic acid | 56472-29-8 | 99.9% | 50 MG
SDP
Catalog No. 5000339683
Encompass_Preferred
(Rac)-Idroxioleic acid is a synthetic oleic acid (OA) derivative that binds to the plasma membrane and alters lipid organization. It has an anti-tumor effect, impairing Jurkat cell growth and inducing proteolytic cleavage of PARP, a molecular marker of apoptosis. In vivo, it significantly inhibits tumor growth in nude mice infected with Jurkat cells.
  • Synthetic oleic acid derivative
  • Alters lipid organization
  • Has anti-tumor effect
  • Impairs Jurkat cell growth in vitro
  • Induces proteolytic cleavage of PARP
  • Inhibits tumor growth in vivo
Catalog No. 50-003-39683 Supplier Medchemexpress LLC Supplier No. HY12946750MG
(Rac)-Idroxioleic acid is a synthetic oleic acid (OA) derivative that binds to the plasma membrane and alters lipid organization. It has an anti-tumor effect, impairing Jurkat cell growth and inducing proteolytic cleavage of PARP, a molecular marker of apoptosis. In vivo, it significantly inhibits tumor growth in nude mice infected with Jurkat cells.
  • Synthetic oleic acid derivative
  • Alters lipid organization
  • Has anti-tumor effect
  • Impairs Jurkat cell growth in vitro
  • Induces proteolytic cleavage of PARP
  • Inhibits tumor growth in vivo