Learn More
Apexbio Technology LLC (Rac)-Indoximod 26988-72-7 50mg
SDP
Catalog No. 5000094900
Encompass_Preferred
(Rac)-Indoximod (CAS 26988-72-7) also known as 1-Methyl-DL-tryptophan is an inhibitor of indoleamine 2 3-dioxygenase (IDO) an enzyme involved in tryptophan catabolism and immune modulation By disrupting IDO-mediated metabolic pathways (Rac)-Indoximod modulates cellular immune responses In human cardiac fibroblast models co-treatment with (Rac)-Indoximod and IFN- reduces -SMA expression and suppresses fibroblast activation This combination also promotes apoptosis via upregulation of IRF-1 Fas and FasL gene expression The compound serves as a valuable tool for studies on IDO regulation and immune cell function
Catalog No. 50-000-94900 Supplier Apexbio Technology LLC Supplier No. A474750
(Rac)-Indoximod (CAS 26988-72-7) also known as 1-Methyl-DL-tryptophan is an inhibitor of indoleamine 2 3-dioxygenase (IDO) an enzyme involved in tryptophan catabolism and immune modulation By disrupting IDO-mediated metabolic pathways (Rac)-Indoximod modulates cellular immune responses In human cardiac fibroblast models co-treatment with (Rac)-Indoximod and IFN- reduces -SMA expression and suppresses fibroblast activation This combination also promotes apoptosis via upregulation of IRF-1 Fas and FasL gene expression The compound serves as a valuable tool for studies on IDO regulation and immune cell function