Learn More
Medchemexpress LLC HY-133897 50mg Medchemexpress, (Rac)-Indoximod CAS:26988-72-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13389750MG
Encompass_Preferred
Medchemexpress, HY-133897 50mg (Rac)-Indoximod CAS:26988-72-7 (Rac)-Indoximod (1-Methyl-DL-tryptophan) is an indoleamine 2,3-dioxygenase (IDO) inhibitor. Co-treatment with IFN-γ and (Rac)-Indoximod markedly reduces the activity of human cardiac myofibroblasts (hCMs) expressing α-SMA and induces apoptosis through up-regulating the IRF-1, Fas, and FasL genes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0304