Learn More
TARGETMOL CHEMICALS INC Rac-JBJ-04-125-02 1MG
SDP
Catalog No. 502876440
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Rac)-JBJ-04-125-02 (JBJ-04-125-02) is effective as a single agent in both in vitro and in vivo models of EGFR-mutant lung cancer. Purity 95.85%
Catalog No. 50-287-6440 Supplier TARGETMOL CHEMICALS INC Supplier No. T88721MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (Rac)-JBJ-04-125-02 (JBJ-04-125-02) is effective as a single agent in both in vitro and in vivo models of EGFR-mutant lung cancer. Purity 95.85%