Learn More
Medchemexpress LLC (Rac)-Juvenile hormone III | 24198-95-6 | MFCD00078044 | 1 MG
SDP
Catalog No. 5000417433
Encompass_Preferred
(Rac)-Juvenile hormone III is identified as a cecropia juvenile hormone and a juvenile hormone agonist. It has been observed to induce test insects to retain larval characteristics even after undergoing their final molt into the adult stage. The substance demonstrates juvenile hormone activity in pupae of *Tenebrio molitor* and in last-instar nymphs of *Oncopeltus fasciatus*. Furthermore, it exhibits morphogenetic activity across various insect assays.
  • Acts as a cecropia juvenile hormone.
  • Functions as a juvenile hormone agonist.
  • Induces the retention of larval characteristics in test insects post-molt.
  • Exhibits juvenile hormone activity in *Tenebrio molitor* pupae.
  • Demonstrates juvenile hormone activity in *Oncopeltus fasciatus* last-instar nymphs.
  • Possesses morphogenetic activity in insect assays.
Catalog No. 50-004-17433 Supplier Medchemexpress LLC Supplier No. HYN72401MG
(Rac)-Juvenile hormone III is identified as a cecropia juvenile hormone and a juvenile hormone agonist. It has been observed to induce test insects to retain larval characteristics even after undergoing their final molt into the adult stage. The substance demonstrates juvenile hormone activity in pupae of *Tenebrio molitor* and in last-instar nymphs of *Oncopeltus fasciatus*. Furthermore, it exhibits morphogenetic activity across various insect assays.
  • Acts as a cecropia juvenile hormone.
  • Functions as a juvenile hormone agonist.
  • Induces the retention of larval characteristics in test insects post-molt.
  • Exhibits juvenile hormone activity in *Tenebrio molitor* pupae.
  • Demonstrates juvenile hormone activity in *Oncopeltus fasciatus* last-instar nymphs.
  • Possesses morphogenetic activity in insect assays.