Learn More
Medchemexpress LLC HY-108235B 10mg Medchemexpress, (Rac)-Lanicemine CAS:61890-25-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108235B10MG
Encompass_Preferred
Medchemexpress, HY-108235B 10mg (Rac)-Lanicemine CAS:61890-25-3 (Rac)-Lanicemine ((Rac)-AZD6765) is the racemate of Lanicemine. Lanicemine (AZD6765) is a low-trapping NMDA channel blocker (Ki of 0.56-2.1 μM for NMDA receptor; IC50s of 4-7 μM and 6.4 μM in CHO and Xenopus oocyte cells, respectively). Antidepressant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0732