Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-lanicemine | 61890-25-3 | MFCD00924222 | 198.27 | C13H14N2 | 5 MG
SDP

Catalog No. 5000241986
Encompass_Preferred

Rac-lanicemine is the racemic form of lanicemine (AZD6765), a low-trapping NMDA receptor channel blocker with reported antidepressant effects. Supplied as a pure analytical standard for research use, it is intended for in vitro and preclinical studies of NMDA receptor pharmacology.

  • Racemic NMDA receptor channel blocker
  • Reported antidepressant activity in preclinical studies
  • Suitable as an analytical standard for in vitro research
  • Molecular formula C13H14N2
  • Molecular weight 198.27
  • Supplied as a pure form
  • Available in research-scale 5 mg quantity

Catalog No. 50-002-41986 Supplier Medchemexpress LLC Supplier No. HY108235B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rac-lanicemine is the racemic form of lanicemine (AZD6765), a low-trapping NMDA receptor channel blocker with reported antidepressant effects. Supplied as a pure analytical standard for research use, it is intended for in vitro and preclinical studies of NMDA receptor pharmacology.

  • Racemic NMDA receptor channel blocker
  • Reported antidepressant activity in preclinical studies
  • Suitable as an analytical standard for in vitro research
  • Molecular formula C13H14N2
  • Molecular weight 198.27
  • Supplied as a pure form
  • Available in research-scale 5 mg quantity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.