Learn More
Medchemexpress LLC (Rac)-Lisaftoclax | 2651980-10-6 | 98.22% | 1 MG
SDP
Catalog No. 5000339552
Encompass_Preferred
(Rac)-Lisaftoclax ((Rac)-APG-2575) is a Bcl-2 inhibitor that can be used for hematologic malignancy research. This product is provided as a powder and should be stored at -20°C for up to 3 years or 4°C for 2 years for optimal stability. Short-term shipping at room temperature is acceptable if the duration is less than 2 weeks. The product container should be kept tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition.
  • Bcl-2 inhibitor for hematologic malignancy research
  • Provided as a high-purity powder
  • Long-term storage stability at -20°C or 4°C
  • Shipped safely at room temperature for durations under two weeks
  • Requires storage in a sealed container in a cool, well-ventilated area
Catalog No. 50-003-39552 Supplier Medchemexpress LLC Supplier No. HY129179B1MG
(Rac)-Lisaftoclax ((Rac)-APG-2575) is a Bcl-2 inhibitor that can be used for hematologic malignancy research. This product is provided as a powder and should be stored at -20°C for up to 3 years or 4°C for 2 years for optimal stability. Short-term shipping at room temperature is acceptable if the duration is less than 2 weeks. The product container should be kept tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition.
  • Bcl-2 inhibitor for hematologic malignancy research
  • Provided as a high-purity powder
  • Long-term storage stability at -20°C or 4°C
  • Shipped safely at room temperature for durations under two weeks
  • Requires storage in a sealed container in a cool, well-ventilated area