Learn More
Medchemexpress LLC HY-110155A 5mg Medchemexpress, (Rac)-LM11A-31 (dihydrochloride) CAS:1214672-15-7 Purity:>98%
SDP
Catalog No. 501968759
Encompass_Preferred
Medchemexpress, HY-110155A 5mg (Rac)-LM11A-31 (diHCl) CAS:1214672-15-7 (Rac)-LM11A-31 dihydrochloride is an isomer of LM11A-31 dihydrochloride. LM11A-31 dihydrochloride, a p75NTR (neurotrophin receptor p75) Ligand, is a potent proNGF (nerve growth factor) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8759 Supplier Medchemexpress LLC Supplier No. HY110155A5MG
Medchemexpress, HY-110155A 5mg (Rac)-LM11A-31 (diHCl) CAS:1214672-15-7 (Rac)-LM11A-31 dihydrochloride is an isomer of LM11A-31 dihydrochloride. LM11A-31 dihydrochloride, a p75NTR (neurotrophin receptor p75) Ligand, is a potent proNGF (nerve growth factor) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.