Learn More
Medchemexpress LLC (Rac)-lunresertib | 2719749-28-5 | 99.7% | C18H20N4O2 | 100 MG
SDP
Catalog No. 5000355265
Encompass_Preferred
(Rac)-lunresertib, also known as (Rac)-RP-6306, is a potent membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) inhibitor with an IC50 of less than 10 nM. This compound exhibits significant anticancer effects.
  • Potent Myt1 inhibitor
  • Inhibits PKMYT1 with an IC50 of <10 nM
  • Exhibits anticancer effects
  • Solid, off-white to yellow appearance
  • Molecular weight of 324.38
Catalog No. 50-003-55265 Supplier Medchemexpress LLC Supplier No. HY145817100MG
(Rac)-lunresertib, also known as (Rac)-RP-6306, is a potent membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) inhibitor with an IC50 of less than 10 nM. This compound exhibits significant anticancer effects.
  • Potent Myt1 inhibitor
  • Inhibits PKMYT1 with an IC50 of <10 nM
  • Exhibits anticancer effects
  • Solid, off-white to yellow appearance
  • Molecular weight of 324.38