Learn More
Medchemexpress LLC (Rac)-lunresertib | 2719749-28-5 | 99.7% | C18H20N4O2 | 50 MG
SDP
Catalog No. 5000355136
Encompass_Preferred
(Rac)-lunresertib (compound 181) is a potent membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) (Gene name PKMYT1) inhibitor with an IC50 of <10 nM. It demonstrates anticancer effects.
  • Potent Myt1 inhibitor
  • Exhibits anticancer effects
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-55136 Supplier Medchemexpress LLC Supplier No. HY14581750MG
(Rac)-lunresertib (compound 181) is a potent membrane-associated tyrosine and threonine-specific cdc2-inhibitory kinase (Myt1) (Gene name PKMYT1) inhibitor with an IC50 of <10 nM. It demonstrates anticancer effects.
  • Potent Myt1 inhibitor
  • Exhibits anticancer effects
  • Soluble in DMSO (100 mg/mL)