Learn More
Medchemexpress LLC Rac-lunresertib | 2719749-28-5 | MFCD34645920 | 99.7% | 324.38 | C18H20N4O2 | 5 MG
SDP
Catalog No. 5000221302
Encompass_Preferred
(Rac)-lunresertib is a racemic, small-molecule inhibitor of the membrane-associated tyrosine- and threonine-specific kinase Myt1 (PKMYT1). It is used as a research reagent in preclinical studies of cell-cycle regulation and anticancer activity and is supplied as a solid or solution with high reported purity. Molecular formula C18H20N4O2; molecular weight 324.38 g/mol.
  • Potent Myt1 inhibition with reported IC50 <10 nM.
  • Suitable for in vitro and preclinical cell-cycle and oncology research.
  • High reported purity (99.72%).
  • Available as an off-white to yellow solid and as solution formulations.
  • Racemic small molecule, convenient handling and storage for lab use.
Catalog No. 50-002-21302 Supplier Medchemexpress LLC Supplier No. HY1458175MG
(Rac)-lunresertib is a racemic, small-molecule inhibitor of the membrane-associated tyrosine- and threonine-specific kinase Myt1 (PKMYT1). It is used as a research reagent in preclinical studies of cell-cycle regulation and anticancer activity and is supplied as a solid or solution with high reported purity. Molecular formula C18H20N4O2; molecular weight 324.38 g/mol.
  • Potent Myt1 inhibition with reported IC50 <10 nM.
  • Suitable for in vitro and preclinical cell-cycle and oncology research.
  • High reported purity (99.72%).
  • Available as an off-white to yellow solid and as solution formulations.
  • Racemic small molecule, convenient handling and storage for lab use.