Learn More
Medchemexpress LLC (Rac)-lunresertib | 2719749-28-5 | MFCD34645920 | 99.7% | C18H20N4O2 | 10MG
SDP
Catalog No. 5000258452
Encompass_Preferred
Lunresertib (racemic) is an orally bioavailable small-molecule inhibitor of PKMYT1 (Myt1) used in preclinical research to study cell-cycle regulation and anticancer activity. It exhibits potent kinase inhibition and is supplied at high analytical purity for laboratory experiments.
  • Potent PKMYT1 inhibitor with IC50 <10 nM.
  • Orally bioavailable small-molecule suitable for cellular and biochemical studies.
  • High analytical purity (about 99.7%).
  • Molecular formula C18H20N4O2 and molecular weight 324.38 g/mol.
  • Supplied in small research pack sizes for laboratory use.
  • Useful for cell-cycle and anticancer mechanism studies.
Catalog No. 50-002-58452 Supplier Medchemexpress LLC Supplier No. HY14581710MG
Lunresertib (racemic) is an orally bioavailable small-molecule inhibitor of PKMYT1 (Myt1) used in preclinical research to study cell-cycle regulation and anticancer activity. It exhibits potent kinase inhibition and is supplied at high analytical purity for laboratory experiments.
  • Potent PKMYT1 inhibitor with IC50 <10 nM.
  • Orally bioavailable small-molecule suitable for cellular and biochemical studies.
  • High analytical purity (about 99.7%).
  • Molecular formula C18H20N4O2 and molecular weight 324.38 g/mol.
  • Supplied in small research pack sizes for laboratory use.
  • Useful for cell-cycle and anticancer mechanism studies.