Learn More
Medchemexpress LLC Rac-Lys-SMCC-DM1 | 00-00-0 | 98.2% | 1103.71 g·mol⁻¹ | C53H75ClN6O15S | 10 MG
SDP
Catalog No. 5000245902
Encompass_Preferred
(Rac)-Lys-SMCC-DM1 is a racemic linker-payload component used in antibody-drug conjugate research. It contains the DM1 cytotoxic payload attached via an SMCC linker to a lysine residue and is commonly used as a reference metabolite, conjugation reagent, and analytical standard in ADC development.
  • Provides a lysine-linked SMCC-DM1 conjugate suitable for ADC research.
  • Serves as an analytical standard for metabolism and stability studies.
  • Enables evaluation of conjugation chemistries and payload release.
  • High supplied purity supports reproducible experimental results.
  • Stable under recommended storage conditions for short- and long-term use.
Catalog No. 50-002-45902 Supplier Medchemexpress LLC Supplier No. HY101982A10MG
(Rac)-Lys-SMCC-DM1 is a racemic linker-payload component used in antibody-drug conjugate research. It contains the DM1 cytotoxic payload attached via an SMCC linker to a lysine residue and is commonly used as a reference metabolite, conjugation reagent, and analytical standard in ADC development.
  • Provides a lysine-linked SMCC-DM1 conjugate suitable for ADC research.
  • Serves as an analytical standard for metabolism and stability studies.
  • Enables evaluation of conjugation chemistries and payload release.
  • High supplied purity supports reproducible experimental results.
  • Stable under recommended storage conditions for short- and long-term use.