Learn More
Medchemexpress LLC HY-101982A 1mg , (Rac)-Lys-SMCC-DM1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101982A1MG
Encompass_Preferred
HY-101982A 1mg (Rac)-Lys-SMCC-DM1 CAS: (Rac)-Lys-SMCC-DM1 ((Rac)-Lys-Nε-MCC-DM1) is the racemate of Lys-SMCC-DM1 (HY-101982). Lys-SMCC-DM1 is a linker-payload component that has the potential to inhibit tubulin polymerization. Lys-SMCC-DM1 is the active metabolite of T-DM1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1253