Learn More
Medchemexpress LLC (Rac)-Lys-SMCC-DM1 ((Rac)-Lys-Nε-MCC-DM1) | 00-00-0 | 98.2% | 1103.71 g/mol | C53H75ClN6O15S | 5 MG
SDP
Catalog No. 5000247979
Encompass_Preferred
(Rac)-Lys-SMCC-DM1 is the racemic form of Lys-SMCC-DM1, a linker-payload component used in antibody-drug conjugate research. It acts as a tubulin polymerization inhibitor and is identified as the active metabolite of T-DM1. Supplied for research use only.
  • Racemate of Lys-SMCC-DM1
  • Used for antibody-drug conjugate development and evaluation
  • Inhibits tubulin polymerization
  • High purity suitable for analytical and preparative work
  • Available in multiple small-scale laboratory pack sizes
  • Store at 4°C under nitrogen; in solvent store at -80°C for long term
Catalog No. 50-002-47979 Supplier Medchemexpress LLC Supplier No. HY101982A5MG
(Rac)-Lys-SMCC-DM1 is the racemic form of Lys-SMCC-DM1, a linker-payload component used in antibody-drug conjugate research. It acts as a tubulin polymerization inhibitor and is identified as the active metabolite of T-DM1. Supplied for research use only.
  • Racemate of Lys-SMCC-DM1
  • Used for antibody-drug conjugate development and evaluation
  • Inhibits tubulin polymerization
  • High purity suitable for analytical and preparative work
  • Available in multiple small-scale laboratory pack sizes
  • Store at 4°C under nitrogen; in solvent store at -80°C for long term