Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-MEM 1003 | 165187-25-7 | 99.6% | 1 ML
SDP

Catalog No. 5000232803
Encompass_Preferred

(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist used in Alzheimer's disease research. It is provided for in vitro research as either solid powder (5-100 MG) or as a ready-to-use 10 mM solution in DMSO (1 ML).

  • Racemate of MEM 1003 for research applications.
  • Potent L-type Ca2+ channel antagonist.
  • Available as 10 mM solution in DMSO (1 ML) and as solid (5-100 MG).
  • High purity (≈99.6%).
  • Molecular weight 432.90 g/mol; formula C22H25ClN2O5.
  • Intended for in vitro research use only.

Catalog No. 50-002-32803 Supplier Medchemexpress LLC Supplier No. HY12160410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist used in Alzheimer's disease research. It is provided for in vitro research as either solid powder (5-100 MG) or as a ready-to-use 10 mM solution in DMSO (1 ML).

  • Racemate of MEM 1003 for research applications.
  • Potent L-type Ca2+ channel antagonist.
  • Available as 10 mM solution in DMSO (1 ML) and as solid (5-100 MG).
  • High purity (≈99.6%).
  • Molecular weight 432.90 g/mol; formula C22H25ClN2O5.
  • Intended for in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.