Learn More
Medchemexpress LLC Rac-mem 1003 | 165187-25-7 | 99.6% | 100 MG
SDP
Catalog No. 5000232946
Encompass_Preferred
(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine compound that antagonizes L-type Ca2+ channels and is used in preclinical research into neurodegenerative disorders such as Alzheimer's disease. The compound is supplied as a solid powder with chemical formula C22H25ClN2O5 and a molecular weight of 432.90 g/mol.
  • Racemic dihydropyridine L-type calcium channel antagonist.
  • High purity solid suitable for research (99.6%).
  • Molecular formula C22H25ClN2O5; molecular weight 432.90 g/mol.
  • Available in small research quantities, including 100 MG solid packages.
Catalog No. 50-002-32946 Supplier Medchemexpress LLC Supplier No. HY121604100MG
(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine compound that antagonizes L-type Ca2+ channels and is used in preclinical research into neurodegenerative disorders such as Alzheimer's disease. The compound is supplied as a solid powder with chemical formula C22H25ClN2O5 and a molecular weight of 432.90 g/mol.
  • Racemic dihydropyridine L-type calcium channel antagonist.
  • High purity solid suitable for research (99.6%).
  • Molecular formula C22H25ClN2O5; molecular weight 432.90 g/mol.
  • Available in small research quantities, including 100 MG solid packages.