Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-MEM 1003 | 165187-25-7 | 99.6% | 25 MG
SDP

Catalog No. 5000232478
Encompass_Preferred

(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist investigated for Alzheimer's disease research. It is supplied as a light yellow powder for research use only.

  • Racemate composition and dihydropyridine class.
  • Molecular formula C22H25ClN2O5.
  • Molecular weight 432.90.
  • Purity 99.6% (as supplied).
  • Physical form: light yellow to yellow solid.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).
  • Available in multiple small-quantity packages, including 25 mg.

Catalog No. 50-002-32478 Supplier Medchemexpress LLC Supplier No. HY12160425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist investigated for Alzheimer's disease research. It is supplied as a light yellow powder for research use only.

  • Racemate composition and dihydropyridine class.
  • Molecular formula C22H25ClN2O5.
  • Molecular weight 432.90.
  • Purity 99.6% (as supplied).
  • Physical form: light yellow to yellow solid.
  • Storage: powder at -20°C (3 years) or 4°C (2 years).
  • Available in multiple small-quantity packages, including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.