Learn More
Medchemexpress LLC (rac)-Metanephrine-d3 hydrochloride | 1215507-88-2 | 97.2% | 236.71 | 5 MG
SDP
Catalog No. 5000452333
Encompass_Preferred
(rac)-Metanephrine-d3 (hydrochloride) is the deuterium labeled (rac)-Metanephrine hydrochloride. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52333 Supplier Medchemexpress LLC Supplier No. HYW012206S5MG
(rac)-Metanephrine-d3 (hydrochloride) is the deuterium labeled (rac)-Metanephrine hydrochloride. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.