Learn More
Medchemexpress LLC (Rac)-minzasolmin | 1802518-63-3 | 99.9% | 425.59 | C23H31N5OS | 5MG
SDP
Catalog No. 5000237236
Encompass_Preferred
(Rac)-Minzasolmin is a racemic small-molecule inhibitor of α-synuclein misfolding with oral bioavailability and blood-brain barrier penetration. It modulates early-stage α-synuclein aggregation and is used in preclinical in vitro and in vivo studies to reduce pathological α-synuclein accumulation in models of Parkinson's disease and related synucleinopathies.
  • High purity, typically reported as 99.9%.
  • Blood-brain barrier permeable small molecule.
  • Racemic composition for preclinical comparison of enantiomeric activity.
  • Molecular formula C23H31N5OS and molecular weight 425.59 g·mol⁻¹.
  • Suitable for both in vitro assays and in vivo model studies.
Catalog No. 50-002-37236 Supplier Medchemexpress LLC Supplier No. HY1252875MG
(Rac)-Minzasolmin is a racemic small-molecule inhibitor of α-synuclein misfolding with oral bioavailability and blood-brain barrier penetration. It modulates early-stage α-synuclein aggregation and is used in preclinical in vitro and in vivo studies to reduce pathological α-synuclein accumulation in models of Parkinson's disease and related synucleinopathies.
  • High purity, typically reported as 99.9%.
  • Blood-brain barrier permeable small molecule.
  • Racemic composition for preclinical comparison of enantiomeric activity.
  • Molecular formula C23H31N5OS and molecular weight 425.59 g·mol⁻¹.
  • Suitable for both in vitro assays and in vivo model studies.