Learn More
TARGETMOL CHEMICALS INC Rac-Modipafant 1MG
SDP
Catalog No. 502877593
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (Rac)-Modipafantis an orally available and selective platelet-activating factor receptor (PAFR) antagonist that inhibits PAF-induced aggregation of washed platelets in rabbits, and can be used to study arthritis and ischemia/reperfusion (I/R) in the superior mesenteric artery (SMA) of the rat. Purity 98.09%
Catalog No. 50-287-7593 Supplier TARGETMOL CHEMICALS INC Supplier No. T280811MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. (Rac)-Modipafantis an orally available and selective platelet-activating factor receptor (PAFR) antagonist that inhibits PAF-induced aggregation of washed platelets in rabbits, and can be used to study arthritis and ischemia/reperfusion (I/R) in the superior mesenteric artery (SMA) of the rat. Purity 98.09%