Learn More
Medchemexpress LLC (Rac)-Modipafant (UK-74505) | 122956-68-7 | 98.15% | 605.09 | 1 MG
SDP
Catalog No. 5000345792
Encompass_Preferred
(Rac)-Modipafant (UK-74505) is an orally active, selective, long-acting irreversible platelet activating factor receptor (PAFR) antagonist. It prevents dengue infection.
  • Orally active
  • Selective
  • Long-acting irreversible platelet activating factor receptor (PAFR) antagonist
  • Prevents dengue infection
Catalog No. 50-003-45792 Supplier Medchemexpress LLC Supplier No. HY10890810MG
(Rac)-Modipafant (UK-74505) is an orally active, selective, long-acting irreversible platelet activating factor receptor (PAFR) antagonist. It prevents dengue infection.
  • Orally active
  • Selective
  • Long-acting irreversible platelet activating factor receptor (PAFR) antagonist
  • Prevents dengue infection