Learn More
Medchemexpress LLC (Rac)-Monepantel-d5 | 2747918-33-6 | 99.4% | 478.42 | 5 MG
SDP
Catalog No. 5000362826
Encompass_Preferred
(Rac)-Monepantel-d5 is deuterium labeled Monepantel, an organic anthelmintic that acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62826 Supplier Medchemexpress LLC Supplier No. HY14774S5MG
(Rac)-Monepantel-d5 is deuterium labeled Monepantel, an organic anthelmintic that acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS