Learn More
Medchemexpress LLC (Rac)-Monepantel sulfone-d5 | 2747918-68-7 | 99.8% | 510.42 | 5 MG
SDP
Catalog No. 5000362827
Encompass_Preferred
(Rac)-Monepantel sulfone-d5 is a deuterium-labeled form of Monepantel, an organic anthelmintic. It acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62827 Supplier Medchemexpress LLC Supplier No. HY14774S15MG
(Rac)-Monepantel sulfone-d5 is a deuterium-labeled form of Monepantel, an organic anthelmintic. It acts as a positive allosteric modulator of a nematode-specific clade of nicotinic acetylcholine receptor (nAChR) subunits.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS