Learn More
Medchemexpress LLC (Rac)-Moxifloxacin ((Rac)-BAY 12-8039 free base) | 354812-41-2 | 99.5% | C21H24FN3O4 | 5 MG
SDP
Catalog No. 5000433816
Encompass_Preferred
Rac-Moxifloxacin is an isoform of Moxifloxacin Hydrochloride, an oral 8-methoxyquinolone antimicrobial. It is used for treating acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Is an oral 8-methoxyquinolone antimicrobial.
  • Used in the treatment of acute bacterial sinusitis.
  • Used in the treatment of acute bacterial exacerbations of chronic bronchitis.
  • Used in the treatment of community-acquired pneumonia.
  • Acts rapidly in vitro, with effects observed within the first 3 hours and complete broth sterilization within 24 hours.
  • Appears to offer a protective effect against macrophage lysis in vitro.
  • Demonstrates efficacy in vivo, leading to longer survival in treated subjects and reduced bacterial overgrowth in spleens and lungs without toxicity.
Catalog No. 50-004-33816 Supplier Medchemexpress LLC Supplier No. HY66011B5MG
Rac-Moxifloxacin is an isoform of Moxifloxacin Hydrochloride, an oral 8-methoxyquinolone antimicrobial. It is used for treating acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
  • Is an oral 8-methoxyquinolone antimicrobial.
  • Used in the treatment of acute bacterial sinusitis.
  • Used in the treatment of acute bacterial exacerbations of chronic bronchitis.
  • Used in the treatment of community-acquired pneumonia.
  • Acts rapidly in vitro, with effects observed within the first 3 hours and complete broth sterilization within 24 hours.
  • Appears to offer a protective effect against macrophage lysis in vitro.
  • Demonstrates efficacy in vivo, leading to longer survival in treated subjects and reduced bacterial overgrowth in spleens and lungs without toxicity.