Learn More
TARGETMOL CHEMICALS INC RAC-NBI-74330 5MG
SDP
Catalog No. 502580724
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. rac-NBI-74330 is an effective and selective CXCR3 antagonist. purity: 97%
Catalog No. 50-258-0724 Supplier TARGETMOL CHEMICALS INC Supplier No. T260355MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. rac-NBI-74330 is an effective and selective CXCR3 antagonist. purity: 97%