Learn More
Medchemexpress LLC (Rac)-NNC 55-0396 | 2517420-92-5 | 96.2% | C30H40Cl2FN3O2 | 5 MG
SDP
Catalog No. 5000358319
Encompass_Preferred
(Rac)-NNC 55-0396 is the racemate of NNC 55-0396 (HY-50722) and is intended for research use only. This product is not classified as a hazardous substance or mixture and is stable under recommended storage conditions.
  • Not a hazardous substance or mixture
  • Stable under recommended storage conditions
  • Recommended storage temperature: -20°C, sealed, away from moisture
  • Detailed safety information is available for handling, first aid, fire fighting, and accidental release
Catalog No. 50-003-58319 Supplier Medchemexpress LLC Supplier No. HY50722A5MG
(Rac)-NNC 55-0396 is the racemate of NNC 55-0396 (HY-50722) and is intended for research use only. This product is not classified as a hazardous substance or mixture and is stable under recommended storage conditions.
  • Not a hazardous substance or mixture
  • Stable under recommended storage conditions
  • Recommended storage temperature: -20°C, sealed, away from moisture
  • Detailed safety information is available for handling, first aid, fire fighting, and accidental release