Learn More
Medchemexpress LLC (Rac)-Norcantharidin | 5442-12-6 | 168.15 | 1 ML
SDP
Catalog No. 5000388911
Encompass_Preferred
(Rac)-Norcantharidin ((Rac)-NCTD) is the racemic form of Norcantharidin. Norcantharidin is a demethylated derivative of Cantharidin and has antitumor activity. This product is supplied as a 10 mM solution in 1 mL of DMSO, making it suitable for various research applications.
  • Demethylated derivative of cantharidin.
  • Exhibits antitumor activity.
  • Supplied as a 10 mM solution.
  • Provided in 1 mL volume.
  • Formulated in DMSO.
Catalog No. 50-003-88911 Supplier Medchemexpress LLC Supplier No. HYN058510MM/1ML
(Rac)-Norcantharidin ((Rac)-NCTD) is the racemic form of Norcantharidin. Norcantharidin is a demethylated derivative of Cantharidin and has antitumor activity. This product is supplied as a 10 mM solution in 1 mL of DMSO, making it suitable for various research applications.
  • Demethylated derivative of cantharidin.
  • Exhibits antitumor activity.
  • Supplied as a 10 mM solution.
  • Provided in 1 mL volume.
  • Formulated in DMSO.