Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-osmi-1 | 2748153-92-4 | 96.5% | 5 MG
SDP

Catalog No. 5000233458
Encompass_Preferred

(Rac)-OSMI-1 is the racemic form of OSMI-1, a cell-permeable small-molecule inhibitor of O-GlcNAc transferase (OGT) used in biochemical and cell-based research to reduce protein O-GlcNAcylation.

  • Cell-permeable OGT inhibitor with reported IC50 ≈ 2.7 μM.
  • Suitable for cell-based assays and biochemical studies.
  • Purity approximately 96.5% (manufacturer-stated).
  • Molecular weight 563.64 g/mol; formula C28H25N3O6S2.
  • Research-use only; not for clinical or human use.

Catalog No. 50-002-33458 Supplier Medchemexpress LLC Supplier No. HY119738A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-OSMI-1 is the racemic form of OSMI-1, a cell-permeable small-molecule inhibitor of O-GlcNAc transferase (OGT) used in biochemical and cell-based research to reduce protein O-GlcNAcylation.

  • Cell-permeable OGT inhibitor with reported IC50 ≈ 2.7 μM.
  • Suitable for cell-based assays and biochemical studies.
  • Purity approximately 96.5% (manufacturer-stated).
  • Molecular weight 563.64 g/mol; formula C28H25N3O6S2.
  • Research-use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.