Learn More
Medchemexpress LLC Rac-osmi-1 | 2748153-92-4 | 96.5% | 50 MG
SDP
Catalog No. 5000233368
Encompass_Preferred
Rac-OSMI-1 is the racemate of OSMI-1, a cell-permeable small-molecule inhibitor of O-GlcNAc transferase (OGT) used in cellular research to reduce protein O-linked N-acetylglucosamine (O-GlcNAc) modification.
  • Inhibits O-GlcNAc transferase with reported IC50 ≈ 2.7 μM.
  • Racemic small molecule for cellular research.
  • Molecular weight 563.64 g/mol.
  • Reported purity 96.5%.
  • Available as a solid in a 50 mg package.
Catalog No. 50-002-33368 Supplier Medchemexpress LLC Supplier No. HY119738A50MG
Rac-OSMI-1 is the racemate of OSMI-1, a cell-permeable small-molecule inhibitor of O-GlcNAc transferase (OGT) used in cellular research to reduce protein O-linked N-acetylglucosamine (O-GlcNAc) modification.
  • Inhibits O-GlcNAc transferase with reported IC50 ≈ 2.7 μM.
  • Racemic small molecule for cellular research.
  • Molecular weight 563.64 g/mol.
  • Reported purity 96.5%.
  • Available as a solid in a 50 mg package.