Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-PF-06256142 | 1609580-97-3 | 99.6% | 1 ML
SDP

Catalog No. 5000233421
Encompass_Preferred

The racemic form of PF-06256142 is a D1 dopamine receptor agonist supplied as a 10 mM solution in DMSO for research use. It is intended for in vitro and in vivo pharmacology studies of D1 receptor signaling.

  • Racemic D1 receptor agonist with EC50 107 nM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research applications.
  • Molecular weight 356.38 g/mol; formula C21H16N4O2.
  • Soluble in DMSO (25 mg/mL) and compatible with multiple in vivo vehicles.
  • Stable under recommended storage conditions for powder and solution.

Catalog No. 50-002-33421 Supplier Medchemexpress LLC Supplier No. HY119943A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The racemic form of PF-06256142 is a D1 dopamine receptor agonist supplied as a 10 mM solution in DMSO for research use. It is intended for in vitro and in vivo pharmacology studies of D1 receptor signaling.

  • Racemic D1 receptor agonist with EC50 107 nM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • High purity suitable for research applications.
  • Molecular weight 356.38 g/mol; formula C21H16N4O2.
  • Soluble in DMSO (25 mg/mL) and compatible with multiple in vivo vehicles.
  • Stable under recommended storage conditions for powder and solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.