Learn More
Medchemexpress LLC HY-108697A 10mg Medchemexpress, (Rac)-PT2399 CAS:1672662-07-5 Purity:>98%
SDP
Catalog No. 501968610
Encompass_Preferred
Medchemexpress, HY-108697A 10mg (Rac)-PT2399 CAS:1672662-07-5 (Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8610 Supplier Medchemexpress LLC Supplier No. HY108697A10MG
Medchemexpress, HY-108697A 10mg (Rac)-PT2399 CAS:1672662-07-5 (Rac)-PT2399 (Compound 10e), the racemate of PT2399, acts as a potent and specific hypoxia-inducible factor 2a (HIF-2α) inhibitor with an IC50 of 0.01 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.