Learn More
Medchemexpress LLC (Rac)-Reparixin | 957407-64-6 | C14H21NO3S | 2 MG
SDP
Catalog No. 5000373089
Encompass_Preferred
(Rac)-Reparixin is an isomer of Reparixin and serves as an experimental control. It is known as a non-competitive allosteric inhibitor, effectively targeting the chemokine receptors CXCR1 and CXCR2 with IC50s of 1 nM and 100 nM, respectively.
  • Isomer of Reparixin
  • Used as an experimental control
  • Non-competitive allosteric inhibitor
  • Targets chemokine receptors CXCR1 and CXCR2
  • Relevant for research in inflammation/immunology, endocrinology, and cancer
Catalog No. 50-003-73089 Supplier Medchemexpress LLC Supplier No. HY15251A2MG
(Rac)-Reparixin is an isomer of Reparixin and serves as an experimental control. It is known as a non-competitive allosteric inhibitor, effectively targeting the chemokine receptors CXCR1 and CXCR2 with IC50s of 1 nM and 100 nM, respectively.
  • Isomer of Reparixin
  • Used as an experimental control
  • Non-competitive allosteric inhibitor
  • Targets chemokine receptors CXCR1 and CXCR2
  • Relevant for research in inflammation/immunology, endocrinology, and cancer