Learn More
Medchemexpress LLC (Rac)-Rhododendrol | 69617-84-1 | 99.76% | 166.22 | 25 MG
SDP
Catalog No. 5000348051
Encompass_Preferred
(Rac)-Rhododendrol, also known as (Rac)-Betuligenol, is an aromatic compound that exhibits pro-oxidant activity and may be useful in suppressing liver diseases. It has been noted to be toxic to melanocytes, potentially leading to cell death. Its metabolite, RD-quinone, is cytotoxic and can cause enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins.
  • Aromatic compound with pro-oxidant activity
  • May be useful in suppressing liver diseases
  • Can be toxic to melanocytes, leading to cell death
  • Metabolite RD-quinone causes enzyme inactivation and endoplasmic reticulum stress
  • Derived melanin exhibits potent pro-oxidant activity
Catalog No. 50-003-48051 Supplier Medchemexpress LLC Supplier No. 25MGHY136913
(Rac)-Rhododendrol, also known as (Rac)-Betuligenol, is an aromatic compound that exhibits pro-oxidant activity and may be useful in suppressing liver diseases. It has been noted to be toxic to melanocytes, potentially leading to cell death. Its metabolite, RD-quinone, is cytotoxic and can cause enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins.
  • Aromatic compound with pro-oxidant activity
  • May be useful in suppressing liver diseases
  • Can be toxic to melanocytes, leading to cell death
  • Metabolite RD-quinone causes enzyme inactivation and endoplasmic reticulum stress
  • Derived melanin exhibits potent pro-oxidant activity