Learn More
Medchemexpress LLC (Rac)-Rhododendrol | 69617-84-1 | 99.8% | 166.22 | 50 MG
SDP
Catalog No. 5000350283
Encompass_Preferred
(Rac)-Rhododendrol is an aromatic compound with pro-oxidant activity, potentially useful in suppressing liver diseases. It can be toxic to melanocytes, leading to cell death. Its metabolite, RD-quinone, is cytotoxic, causing enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins. Additionally, (Rac)-Rhododendrol-derived melanin exhibits potent pro-oxidant activity and may cause oxidative stress.
  • Aromatic compound with pro-oxidant activity
  • May be useful in suppressing liver diseases
  • Can be toxic to melanocytes
  • Metabolite RD-quinone is cytotoxic
  • Exhibits in vitro cell growth inhibitory activity against K562 cell line
  • Soluble in DMSO at 200 mg/mL
Catalog No. 50-003-50283 Supplier Medchemexpress LLC Supplier No. HY13691350MG
(Rac)-Rhododendrol is an aromatic compound with pro-oxidant activity, potentially useful in suppressing liver diseases. It can be toxic to melanocytes, leading to cell death. Its metabolite, RD-quinone, is cytotoxic, causing enzyme inactivation and endoplasmic reticulum stress by binding to thiol proteins. Additionally, (Rac)-Rhododendrol-derived melanin exhibits potent pro-oxidant activity and may cause oxidative stress.
  • Aromatic compound with pro-oxidant activity
  • May be useful in suppressing liver diseases
  • Can be toxic to melanocytes
  • Metabolite RD-quinone is cytotoxic
  • Exhibits in vitro cell growth inhibitory activity against K562 cell line
  • Soluble in DMSO at 200 mg/mL