Learn More
Medchemexpress LLC (Rac)-Rotigotine-d3 hydrochloride | 1215846-20-0 | C19H23D3ClNOS | 1 MG
SDP
Catalog No. 5000376698
Encompass_Preferred
(Rac)-Rotigotine-d3 hydrochloride is a deuterium labeled (Rac)-Rotigotine (hydrochloride), which is a racemate of Rotigotine. Rotigotine acts as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Ki values for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor ranging from 0.71 nM to 83 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as a full agonist of dopamine receptors.
  • Acts as a partial agonist of the 5-HT1A receptor.
  • Antagonizes the α2B-adrenergic receptor.
Catalog No. 50-003-76698 Supplier Medchemexpress LLC Supplier No. HY15394S11MG
(Rac)-Rotigotine-d3 hydrochloride is a deuterium labeled (Rac)-Rotigotine (hydrochloride), which is a racemate of Rotigotine. Rotigotine acts as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Ki values for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor ranging from 0.71 nM to 83 nM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as a full agonist of dopamine receptors.
  • Acts as a partial agonist of the 5-HT1A receptor.
  • Antagonizes the α2B-adrenergic receptor.