Learn More
Medchemexpress LLC (Rac)-Rotigotine hydrochloride | 102120-99-0 | C19H26ClNOS | 1 ML
SDP
Catalog No. 5000376324
Encompass_Preferred
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.
  • Full agonist of dopamine receptor.
  • Partial agonist of the 5-HT1A receptor.
  • Antagonist of the α2B-adrenergic receptor.
  • Exhibits selectivity for D3 receptors.
  • Protects dopaminergic neurons against rotenone-induced cell death.
  • Inhibits ROS production by rotenone.
Catalog No. 50-003-76324 Supplier Medchemexpress LLC Supplier No. HY1539410MM/1ML
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor.
  • Full agonist of dopamine receptor.
  • Partial agonist of the 5-HT1A receptor.
  • Antagonist of the α2B-adrenergic receptor.
  • Exhibits selectivity for D3 receptors.
  • Protects dopaminergic neurons against rotenone-induced cell death.
  • Inhibits ROS production by rotenone.