Learn More
Medchemexpress LLC (Rac)-Rotigotine hydrochloride | 102120-99-0 | C19H26ClNOS | 5 MG
SDP
Catalog No. 5000376514
Encompass_Preferred
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine, functioning as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It is primarily utilized in research contexts, especially for studies related to Parkinson's disease.
  • Functions as a full agonist at dopamine receptors
  • Acts as a partial agonist at the 5-HT1A receptor
  • Serves as an antagonist at the alpha-2B adrenergic receptor
  • Shows selectivity for D3 receptors over D2, D4, D5, and D1 receptors
  • Provides protection for dopaminergic neurons against MPP+ toxicity
  • Significantly inhibits ROS production caused by rotenone
Catalog No. 50-003-76514 Supplier Medchemexpress LLC Supplier No. HY153945MG
(Rac)-Rotigotine hydrochloride is a racemate of Rotigotine, functioning as a full agonist of the dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor. It is primarily utilized in research contexts, especially for studies related to Parkinson's disease.
  • Functions as a full agonist at dopamine receptors
  • Acts as a partial agonist at the 5-HT1A receptor
  • Serves as an antagonist at the alpha-2B adrenergic receptor
  • Shows selectivity for D3 receptors over D2, D4, D5, and D1 receptors
  • Provides protection for dopaminergic neurons against MPP+ toxicity
  • Significantly inhibits ROS production caused by rotenone