Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-ruxolitinib-d9 | 2469553-67-9 | 99.4% | 315.42 | C17H9D9N6 | 1 MG
SDP

Catalog No. 5000210666
Encompass_Preferred

(Rac)-Ruxolitinib-d9 is the deuterium-labeled racemic form of ruxolitinib (INCB18424), a JAK1/2 inhibitor supplied for research use as an analytical standard or tracer in biochemical and pharmacokinetic studies.

  • Deuterium-labeled tracer for isotope labeling and mass spectrometry applications.
  • High purity suitable for analytical work (99.4%).
  • Provided as a solid, white to yellow, for ease of handling and storage.
  • Available in small research sizes suitable for standards (1 mg and 5 mg).
  • Documented identifiers: CAS number 2469553-67-9; molecular formula C17H9D9N6; molecular weight 315.42.
  • Storage guidance: long-term at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month (stored under nitrogen).

Catalog No. 50-002-10666 Supplier Medchemexpress LLC Supplier No. HYW062703S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Ruxolitinib-d9 is the deuterium-labeled racemic form of ruxolitinib (INCB18424), a JAK1/2 inhibitor supplied for research use as an analytical standard or tracer in biochemical and pharmacokinetic studies.

  • Deuterium-labeled tracer for isotope labeling and mass spectrometry applications.
  • High purity suitable for analytical work (99.4%).
  • Provided as a solid, white to yellow, for ease of handling and storage.
  • Available in small research sizes suitable for standards (1 mg and 5 mg).
  • Documented identifiers: CAS number 2469553-67-9; molecular formula C17H9D9N6; molecular weight 315.42.
  • Storage guidance: long-term at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month (stored under nitrogen).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.