Learn More
Medchemexpress LLC (Rac)-Salvianic acid A sodium ((rac)-Danshensu sodium) | 67920-52-9 | 98.0% | 220.15 | 200 MG
SDP
Catalog No. 5000427504
Encompass_Preferred
(Rac)-Salvianic acid A sodium is the racemic form of Salvianic acid A, a phenolic compound known for inducing Nrf2/HO-1 activation and inhibiting the NF-κB pathway. It also activates the mitochondrial antioxidant defense system. This compound demonstrates anti-inflammatory, antioxidant, and anti-apoptotic properties.
  • Exhibits anti-inflammatory effects.
  • Acts as an antioxidant.
  • Demonstrates anti-apoptotic properties.
  • Induces Nrf2/HO-1 activation.
  • Inhibits the NF-κB pathway.
  • Activates the mitochondrial antioxidant defense system.
Catalog No. 50-004-27504 Supplier Medchemexpress LLC Supplier No. HYN0106200MG
(Rac)-Salvianic acid A sodium is the racemic form of Salvianic acid A, a phenolic compound known for inducing Nrf2/HO-1 activation and inhibiting the NF-κB pathway. It also activates the mitochondrial antioxidant defense system. This compound demonstrates anti-inflammatory, antioxidant, and anti-apoptotic properties.
  • Exhibits anti-inflammatory effects.
  • Acts as an antioxidant.
  • Demonstrates anti-apoptotic properties.
  • Induces Nrf2/HO-1 activation.
  • Inhibits the NF-κB pathway.
  • Activates the mitochondrial antioxidant defense system.