Learn More
TARGETMOL CHEMICALS INC Rac-SAR131675 25MG
SDP
Catalog No. 502876744
Encompass_Preferred
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.(Rac)-SAR131675 is an effective and specific VEGFR-3 inhibitor. It inhibited VEGFR-3 tyrosine kinase activity (IC50 20 nmol/L) and VEGFR-3 autophosphorylation (IC50 45 nmol/L) in HEK cells, respectively. SAR131675 is highly specific for VEGFR-3 versus 107 receptors, enzymes, ion channels, and 65 kinases. SAR131675 is a highly specific VEGFR-3-TK inhibitor with significant antitumoral and antimetastatic activities in vivo through inhibition of lymphangiogenesis and TAM invasion. Purity 98.34%
Catalog No. 50-287-6744 Supplier TARGETMOL CHEMICALS INC Supplier No. T369125MG
Also available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.(Rac)-SAR131675 is an effective and specific VEGFR-3 inhibitor. It inhibited VEGFR-3 tyrosine kinase activity (IC50 20 nmol/L) and VEGFR-3 autophosphorylation (IC50 45 nmol/L) in HEK cells, respectively. SAR131675 is highly specific for VEGFR-3 versus 107 receptors, enzymes, ion channels, and 65 kinases. SAR131675 is a highly specific VEGFR-3-TK inhibitor with significant antitumoral and antimetastatic activities in vivo through inhibition of lymphangiogenesis and TAM invasion. Purity 98.34%