Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-SCH 563705 | 2701435-64-3 | 99.8% | 425.48 | C23H27N3O5 | 10 MG
SDP

Catalog No. 5000215463
Encompass_Preferred

(Rac)-SCH 563705 is the racemic form of SCH 563705, a potent, orally active antagonist of CXCR2 and CXCR1 used as a research tool to study chemokine receptor signaling. The compound is supplied as a solid for laboratory research applications.

  • High purity: 99.78%.
  • Molecular formula C23H27N3O5 and molecular weight 425.48.
  • CAS number 2701435-64-3.
  • Soluble in DMSO at 200 mg/mL with ultrasonication.
  • Off-white to light-yellow solid.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Catalog No. 50-002-15463 Supplier Medchemexpress LLC Supplier No. HY10011B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-SCH 563705 is the racemic form of SCH 563705, a potent, orally active antagonist of CXCR2 and CXCR1 used as a research tool to study chemokine receptor signaling. The compound is supplied as a solid for laboratory research applications.

  • High purity: 99.78%.
  • Molecular formula C23H27N3O5 and molecular weight 425.48.
  • CAS number 2701435-64-3.
  • Soluble in DMSO at 200 mg/mL with ultrasonication.
  • Off-white to light-yellow solid.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.