Learn More
Medchemexpress LLC Rac-SCH 563705 | 473728-58-4 | 99.8% | 425.48 | C23H27N3O5 | 50 MG
SDP
Catalog No. 5000215720
Encompass_Preferred
(Rac)-SCH 563705 is the racemic form of a potent, orally active antagonist of chemokine receptors CXCR2 and CXCR1, supplied as a high-purity research compound for in vitro and in vivo studies. It exhibits low-nanomolar receptor potency and is provided with analytical documentation and handling guidance.
Catalog No. 50-002-15720 Supplier Medchemexpress LLC Supplier No. HY10011B50MG
(Rac)-SCH 563705 is the racemic form of a potent, orally active antagonist of chemokine receptors CXCR2 and CXCR1, supplied as a high-purity research compound for in vitro and in vivo studies. It exhibits low-nanomolar receptor potency and is provided with analytical documentation and handling guidance.