Learn More
TARGETMOL CHEMICALS INC RAC-SHIKONIN 50MG
SDP
Catalog No. 502580473
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-Shikonin (Anchusin) is a natural red naphthoquinone pigment has antimicrobial anti-tumor and anti-inflammatory effects; a purified shikonin preparation is widely used for the production of medicinals cosmetics and some food products; shikonin also enters into the antiinflammatory ointment and cream compositions used for the treatment of burns. It can suppress the cell viability adhesion invasion and migratory ability of MGC-803 cells through TLR2- or NF-(kappa)B-mediated pathway. purity: 99%
Catalog No. 50-258-0473 Supplier TARGETMOL CHEMICALS INC Supplier No. T289750MG
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-Shikonin (Anchusin) is a natural red naphthoquinone pigment has antimicrobial anti-tumor and anti-inflammatory effects; a purified shikonin preparation is widely used for the production of medicinals cosmetics and some food products; shikonin also enters into the antiinflammatory ointment and cream compositions used for the treatment of burns. It can suppress the cell viability adhesion invasion and migratory ability of MGC-803 cells through TLR2- or NF-(kappa)B-mediated pathway. purity: 99%