Learn More
Medchemexpress LLC (Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 10 MG
SDP
Catalog No. 5000436142
Encompass_Preferred
(Rac)-sn-Glycerol 3-phosphate is an a-site substrate analogue. It binds to the a-site, strongly inhibiting the reaction rate of indole and nucleophilic indole analogues with E(A-A). It is also a phosphorylated derivative of glycerol produced as an intermediate in the glycolysis pathway. (Rac)-sn-Glycerol 3-phosphate binds to the α subunit of tryptophan synthase, regulating tryptophan synthesis through allosteric inhibition. It provides microorganisms with advantages in nutrient-lacking environments as a phosphorus, carbon, and energy source, and can be used for microorganism cultivation. Additionally, it binds to metal ions, forming insoluble phosphate minerals, and can be used for in situ bioremediation to reduce environmental pollution.
  • Endogenous metabolite
  • A-site substrate analogue
  • Can be used for microorganism cultivation
  • Can be used for in situ bioremediation
Catalog No. 50-004-36142 Supplier Medchemexpress LLC Supplier No. HY113128C10MG
(Rac)-sn-Glycerol 3-phosphate is an a-site substrate analogue. It binds to the a-site, strongly inhibiting the reaction rate of indole and nucleophilic indole analogues with E(A-A). It is also a phosphorylated derivative of glycerol produced as an intermediate in the glycolysis pathway. (Rac)-sn-Glycerol 3-phosphate binds to the α subunit of tryptophan synthase, regulating tryptophan synthesis through allosteric inhibition. It provides microorganisms with advantages in nutrient-lacking environments as a phosphorus, carbon, and energy source, and can be used for microorganism cultivation. Additionally, it binds to metal ions, forming insoluble phosphate minerals, and can be used for in situ bioremediation to reduce environmental pollution.
  • Endogenous metabolite
  • A-site substrate analogue
  • Can be used for microorganism cultivation
  • Can be used for in situ bioremediation