Learn More
Medchemexpress LLC (Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 100 MG
SDP
Catalog No. 5000437194
Encompass_Preferred
(Rac)-sn-Glycerol 3-phosphate is a phosphorylated derivative of glycerol and an intermediate in the glycolysis pathway. It is an α-site substrate analogue that strongly inhibits the reaction rate of indole and nucleophilic indole analogues with E(A-A) when bound to the α-site. This compound binds to the α subunit of tryptophan synthase, allosterically regulating tryptophan synthesis. It can also serve as a phosphorus, carbon, and energy source for microorganisms.
  • Useful for microbial cultivation
  • Binds to metal ions to form insoluble phosphate minerals
  • Utilized for in situ bioremediation to reduce environmental pollution
Catalog No. 50-004-37194 Supplier Medchemexpress LLC Supplier No. HY113128C100MG
(Rac)-sn-Glycerol 3-phosphate is a phosphorylated derivative of glycerol and an intermediate in the glycolysis pathway. It is an α-site substrate analogue that strongly inhibits the reaction rate of indole and nucleophilic indole analogues with E(A-A) when bound to the α-site. This compound binds to the α subunit of tryptophan synthase, allosterically regulating tryptophan synthesis. It can also serve as a phosphorus, carbon, and energy source for microorganisms.
  • Useful for microbial cultivation
  • Binds to metal ions to form insoluble phosphate minerals
  • Utilized for in situ bioremediation to reduce environmental pollution