Learn More
Medchemexpress LLC (Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 25 MG
SDP
Catalog No. 5000439614
Encompass_Preferred
(Rac)-sn-Glycerol 3-phosphate is an α-site substrate analogue and a phosphorylated derivative of glycerol, produced as an intermediate in the glycolysis pathway. It regulates tryptophan synthesis through allosteric inhibition and can be used for cultivating microorganisms by providing phosphorus, carbon, and energy. It also binds to metal ions to form insoluble phosphate minerals, which can be utilized for in situ bioremediation.
  • a-site substrate analogue
  • Endogenous metabolite
  • Appears as a colorless to light yellow liquid
  • Strongly inhibits the reaction of indole and nucleophilic indole analogues
  • Regulates tryptophan synthesis through allosteric inhibition
  • Provides microorganisms with phosphorus, carbon, and energy sources
  • Can be used for in situ bioremediation
Catalog No. 50-004-39614 Supplier Medchemexpress LLC Supplier No. 25MGHY113128C
(Rac)-sn-Glycerol 3-phosphate is an α-site substrate analogue and a phosphorylated derivative of glycerol, produced as an intermediate in the glycolysis pathway. It regulates tryptophan synthesis through allosteric inhibition and can be used for cultivating microorganisms by providing phosphorus, carbon, and energy. It also binds to metal ions to form insoluble phosphate minerals, which can be utilized for in situ bioremediation.
  • a-site substrate analogue
  • Endogenous metabolite
  • Appears as a colorless to light yellow liquid
  • Strongly inhibits the reaction of indole and nucleophilic indole analogues
  • Regulates tryptophan synthesis through allosteric inhibition
  • Provides microorganisms with phosphorus, carbon, and energy sources
  • Can be used for in situ bioremediation